Merck
  • Electron transfer sensitized photodechlorination of surfactant solubilized PCB 138.
Electron transfer sensitized photodechlorination of surfactant solubilized PCB 138.
Chemosphere (2012-11-13)
Linlong Yu, Maryam Izadifard, Gopal Achari, Cooper H Langford
ABSTRACT
Sensitized photodechlorination of polychlorinated biphenyl, PCB 138, in three different surfactant solutions was studied. The sensitizer of choice was leuco-methylene blue, which was produced in situ from methylene blue using either triethylamine or sodium borohydride. Three types of surfactants, anionic (SDS), neutral (TWEEN 80), and cationic (CTAB) at different concentrations were investigated. The neutral and cationic surfactants were found to be more effective than anionic. In each case the surfactant concentration was found to play a significant role in the rate of dechlorination. For different sensitized systems (triethylamine or sodium borohydride), a different product distribution and a different pathway of dechlorination was observed.
Product Description
2,2′,3,4,4′,5′-Hexachlorobiphenyl (IUPAC No. 138), BCR®, certified reference material
SKU
Pack Size
Availability
Price
Quantity
Supelco
PCB No 138, analytical standard