Merck
Total synthesis of (-)-orthodiffenes A and C.
The Journal of organic chemistry (2012-10-12)
Jun Liu, Yi Liu, Xing Zhang, Chaoli Zhang, Yangguang Gao, LinLin Wang, Yuguo Du
ABSTRACT
The efficient and concise synthesis of (-)-orthodiffenes A and C has been accomplished for the first time in eight steps from readily available chiral synthons, D-mannose and D-ethyl lactate. Our work confirmed the complete structure of orthodiffenes A and C, including their absolute stereochemistry. The key steps of our total synthesis involved cis-fused tetrahydrofuran cyclization, one-pot deprotection-lactonization, and intramolecular benzoyl migration according to a biosynthetic hypothesis of orthodiffenes.
Sigma-Aldrich
(−)-Ethyl L-lactate, photoresist grade, ≥99.0%
Sigma-Aldrich
Ethyl lactate, natural, ≥98%, FCC, FG
Sigma-Aldrich
(−)-Ethyl L-lactate, 98%
Sigma-Aldrich
Ethyl lactate, ≥98%, FCC, FG
Sigma-Aldrich
(−)-Ethyl L-lactate, purum, ≥98.0% (sum of enantiomers, GC)