Merck
  • Interaction of local anesthetics with lipid bilayers investigated by ¹H MAS NMR spectroscopy.
Interaction of local anesthetics with lipid bilayers investigated by ¹H MAS NMR spectroscopy.
Biochimica et biophysica acta (2012-07-31)
Nicole Weizenmann, Daniel Huster, Holger A Scheidt
ABSTRACT
The membrane location of the local anesthetics (LA) lidocaine, dibucaine, tetracaine, and procaine hydrochloride as well as their influence on phospholipid bilayers were studied by ³¹P and ¹H magic-angle spinning (MAS) NMR spectroscopy. The ³¹P NMR spectra of the LA/lipid preparations confirmed that the overall bilayer structure of the membrane remained preserved. The relation between the molecular structure of the LAs and their membrane localization and orientation was investigated quantitatively using induced chemical shifts, nuclear Overhauser enhancement spectroscopy, and paramagnetic relaxation rates. All three methods revealed an average location of the aromatic rings of all LAs in the lipid-water interface of the membrane, with small differences between the individual LAs depending on their molecular properties. While lidocaine is placed in the upper chain/glycerol region of the membrane, for dibucaine and procaine the maximum of the distribution are slightly shifted into the glycerol region. Finally for tetracaine the aromatic ring is placed closest to the aqueous phase in the glycerol/headgroup region of the membrane. The hydrophobic side chains of the LA molecules dibucaine and tetracaine were located deeper in the membrane and showed an orientation towards the hydrocarbon core. In contrast the side chains of lidocaine and procaine are oriented towards the aqueous phase.
Product Description
Supelco
Procaine hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Tetracaine hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Procaine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Tetracaine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Tetracaine for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Tetracaine, ≥98% (TLC)
Avanti
16:0 Tempo PC, Avanti Research - A Croda Brand 810606P, powder
Sigma-Aldrich
Tetracaine hydrochloride, ≥99%
Sigma-Aldrich
Procaine hydrochloride, ≥97%