Merck
  • Novel solid self-emulsifying drug delivery system of coenzyme Q₁₀ with improved photochemical and pharmacokinetic behaviors.
Novel solid self-emulsifying drug delivery system of coenzyme Q₁₀ with improved photochemical and pharmacokinetic behaviors.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences (2012-04-14)
Satomi Onoue, Atushi Uchida, Kazuki Kuriyama, Tatsuya Nakamura, Yoshiki Seto, Masashi Kato, Junya Hatanaka, Toshiyuki Tanaka, Hiroyuki Miyoshi, Shizuo Yamada
ABSTRACT
The present study was undertaken to develop a solid self-emulsifying drug delivery system of coenzyme Q(10) (CoQ(10)/s-SEDDS) with high photostability and oral bioavailability. The CoQ(10)/s-SEDDS was prepared by spray-drying an emulsion preconcentrate containing CoQ(10), medium-chain triglyceride, sucrose ester of fatty acid, and hydroxypropyl cellulose, and its physicochemical, photochemical, and pharmacokinetic properties were evaluated. The CoQ(10)/s-SEDDS powder with a diameter of ca. 15 μm was obtained by spray-drying, in which the CoQ(10) was mostly amorphized. The CoQ(10)/s-SEDDS exhibited immediate self-emulsification when introduced to aqueous media under gentle agitation, forming uniform fine droplets with a mean diameter of ca. 280 nm. There was marked generation of reactive oxygen species, in particular superoxide, from CoQ(10) exposed to simulated sunlight (250W/m(2)), suggesting potent photoreactivity. Nano-emulsified solution of CoQ(10) under light exposure underwent photodegradation with 22-fold higher degradation kinetics than crystalline CoQ(10), although the CoQ(10)/s-SEDDS was less photoreactive. After the oral administration of CoQ(10)/s-SEDDS (100 mg-CoQ(10)/kg) in rats, enhanced exposure of CoQ(10) was observed with increases in both C(max) and AUC of ca. 5-fold in comparison with those of orally administered crystalline CoQ(10). From the improved physicochemical and pharmacokinetic data, the s-SEDDS approach upon spray-drying might be a suitable dosage option for enhancing nutraceutical and pharmaceutical values of CoQ(10).
Sigma-Aldrich
Hydroxypropyl cellulose, average Mw ~80,000, average Mn ~10,000, powder, 20 mesh particle size (99% through)
Sigma-Aldrich
Hydroxypropyl cellulose, average Mw ~1,000,000, powder, 20 mesh particle size (99% through)
Sigma-Aldrich
Hydroxypropyl cellulose, average Mw ~100,000, powder, 20 mesh particle size (99% through)
Sigma-Aldrich
Hydroxypropyl cellulose, average Mw ~370,000, powder, 20 mesh particle size (99% through)