Merck
  • Toxicological evaluation of three contaminants of emerging concern by use of the Allium cepa test.
Toxicological evaluation of three contaminants of emerging concern by use of the Allium cepa test.
Mutation research (2012-01-18)
O Herrero, J M Pérez Martín, P Fernández Freire, L Carvajal López, A Peropadre, M J Hazen
ABSTRACT
Di(2-ethylhexyl)phthalate, triclosan and propylparaben are contaminants of emerging concern that have been subjected to extensive toxicological studies, but for which limited information is currently available concerning adverse effects on terrestrial plant systems. The Allium cepa test, which is considered one of the most efficient approaches to assess toxic effects of environmental chemicals, was selected to evaluate the potential risks of these ubiquitous pollutants. Our data demonstrate that all three compounds studied may in some way be considered toxic, but different effects were noted depending on the chemical and the end point analysed. Results derived from the analysis of macroscopic parameters used in testing for general toxicity, revealed that while di(2-ethylhexyl)phthalate had no apparent effects, the other two chemicals inhibited A. cepa root growth in a dose-dependent manner. On the other hand, although all three compounds caused alterations in the mitotic index of root-tip cells, propylparaben was the only one that did not show evidence of genotoxicity in assays for chromosome aberrations and micronuclei. The results of the present study clearly indicate that sensitive plant bioassays are useful and complementary tools to determine environmental impact of contaminants of emerging concern.
Product Description
Propyl parahydroxybenzoate, European Pharmacopoeia (EP) Reference Standard
Supelco
Propylparaben, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Propyl 4-hydroxybenzoate, ≥99%
Sigma-Aldrich
Propyl 4-hydroxybenzoate, tested according to Ph. Eur.
Sigma-Aldrich
Propyl 4-hydroxybenzoate, BioXtra