Merck
  • A laccase of Fomes durissimus MTCC-1173 and its role in the conversion of methylbenzene to benzaldehyde.
A laccase of Fomes durissimus MTCC-1173 and its role in the conversion of methylbenzene to benzaldehyde.
Applied biochemistry and biotechnology (2011-11-15)
R Sahay, R S S Yadav, Sudha Yadava, K D S Yadav
ABSTRACT
A laccase has been purified from the liquid culture growth medium containing bagasse particles of Fomes durissimus. The method involved concentration of the culture filtrate by ultrafiltration and anion exchange chromatography on diethyl aminoethyl cellulose. The sodium dodecyl sulphate-polyacrylamide gel electrophoresis (SDS-PAGE) and native polyacrylamide gel electrophoresis both gave single protein band indicating that the enzyme preparation was pure. The molecular mass of the purified laccase determined from SDS-PAGE analysis was 75 kDa. Using 2,6-dimethoxyphenol as the substrate, the determined K (m) and k (cat) values of the laccase are 182 μM and 0.35 s(-1), respectively, giving a k (cat)/K (m) value of 1.92 × 10(3) M(-1) s(-1). The pH and temperature optimum were 4.0 and 35 °C, respectively. The purified laccase has yellow colour and does not show absorption band around 610 nm found in blue laccases. Moreover, it transformed methylbenzene to benzaldehyde in the absence of mediator molecules, property exhibited by yellow laccases.
Product Description
Supelco
2,6-Dimethoxyphenol, analytical standard
Sigma-Aldrich
2,6-Dimethoxyphenol, natural, ≥96%
Sigma-Aldrich
2,6-Dimethoxyphenol, 99%
Sigma-Aldrich
2,6-Dimethoxyphenol, ≥98%, FG