Merck
  • Development and validation of transferable amide I vibrational frequency maps for peptides.
Development and validation of transferable amide I vibrational frequency maps for peptides.
The journal of physical chemistry. B (2011-03-17)
L Wang, C T Middleton, M T Zanni, J L Skinner
ABSTRACT
Infrared (IR) spectroscopy of the amide I band has been widely utilized for the analysis of peptides and proteins. Theoretical modeling of IR spectra of proteins requires an accurate and efficient description of the amide I frequencies. In this paper, amide I frequency maps for protein backbone and side chain groups are developed from experimental spectra and vibrational lifetimes of N-methylacetamide and acetamide in different solvents. The frequency maps, along with established nearest-neighbor frequency shift and coupling schemes, are then applied to a variety of peptides in aqueous solution and reproduce experimental spectra well. The frequency maps are designed to be transferable to different environments; therefore, they can be used for heterogeneous systems, such as membrane proteins.
Sigma-Aldrich
Acetamide, ~99% (GC)
Sigma-Aldrich
N-Methylacetamide, ≥99%
Sigma-Aldrich
Acetamide, ≥99.0% (GC)
Sigma-Aldrich
Acetamide, sublimed, 99%