Merck
The liquid structure of haloforms CHCl3 and CHBr3.
Journal of physics. Condensed matter : an Institute of Physics journal (2011-03-10)
Szilvia Pothoczki, László Temleitner, Shinji Kohara, Pál Jóvári, László Pusztai
ABSTRACT
New neutron and x-ray diffraction measurements are reported on liquid chloroform, CHCl(3), and bromoform, CHBr(3). Experimental total scattering structure factors have been interpreted by the reverse Monte Carlo method of structural modelling. Partial radial distribution functions, intramolecular bond angle distributions and functions characterizing distance-dependent orientational correlations have been calculated directly from the particle coordinates. It has been found that most of these characteristics of the microscopic structure can be approximated rather well by functions calculated for hard sphere like reference systems. The two liquids show similar features from the point of view of their structure. There are also some distinctive features in terms of orientational correlations: nearest neighbour molecules prefer face-to-face arrangement in chloroform whereas in bromoform, edge-to-face configurations dominate, with a significant occurrence of corner-to-face type correlations.
Sigma-Aldrich
Bromoform, contains 1-3% ethanol as stabilizer, 96%
Supelco
Bromoform, amylene stabilized, analytical standard
Sigma-Aldrich
Bromoform, contains 60-120 ppm 2-methyl-2-butene as stabilizer, 99%
Supelco
Bromoform solution, certified reference material, 5000 μg/mL in methanol