Merck
  • Pyrophosphate-mediated magnetic interactions in Cu(II) coordination complexes.
Pyrophosphate-mediated magnetic interactions in Cu(II) coordination complexes.
Inorganic chemistry (2010-12-01)
Nadia Marino, Oluwatayo F Ikotun, Miguel Julve, Francesc Lloret, Juan Cano, Robert P Doyle
ABSTRACT
The reaction in water of Cu(NO(3))(2)·2.5H(2)O with 2,2'-bipyridine (bipy), 1,10-phenanthroline (phen), or 1,10-phenanthroline-5-amine (phenam), and sodium pyrophosphate (Na(4)P(2)O(7)), at various pHs, afforded three new copper(II)-pyrophosphate complexes, namely, {[Cu(bipy)(cis-H(2)P(2)O(7))](2)}·3H(2)O (1a), {[Cu(phen)(H(2)O)](4)(HP(2)O(7))(2)}(ClO(4))(2)·4H(2)O (2), and {[Cu(2)(phenam)(2)(P(2)O(7))](2)·25H(2)O}(n) (3). A solvent free crystalline phase of 1a was also isolated with formula {[Cu(bipy)(trans-H(2)P(2)O(7))](2)} (1b), which can be regarded as a pseudo-polymorph of 1a. Single crystal X-ray analyses revealed these compounds to have uncommon molecular architectures, with 3 being an unprecedented pyrophosphate-containing two-dimensional (2D) polymer. Compounds 1a/1b and 2 are discrete di- and tetra-nuclear complexes, respectively. The cationic {[Cu(phen)(H(2)O)](4)(HP(2)O(7))(2)}(2+) unit in 2 presents a unique quasi-flat structure, held together by solely in-plane pyrophosphate bridging modes (short O(eq)-P-O(eq) and long O(eq)-P-O-P-O(eq) pathways), a coordination arrangement also not previously reported. A different tetranuclear copper(II)-pyrophosphate arrangement is found in 3, with two classically bridged dimers (O(eq)-P-O(eq) pathway) joined together by auxiliary equatorial-axial μ-O pyrophosphate bridges. Here, the bidimensionality is reached through bridging phenam ligands, which provide further inter-"tetramer" metal-metal connections [(N,N')(eq)-(N'')(ax) pathway], leading to the formation of an expanded covalent network based on the [Cu(2)(phenam)(2)(P(2)O(7))](2) moiety. Variable-temperature magnetic susceptibility measurements on polycrystalline samples of 2 and 3 revealed net antiferromagnetic coupling between metal centers with J(2a) = -7.9(2) cm(-1), J(2b) = -46.9(3) cm(-1), J(2c) = 0 cm(-1) in 2 (H = -J(2a)[S(Cu(1))·S(Cu(2)) + S(Cu(1a))·S(Cu(2a))] - J(2b)[S(Cu(1))·S(Cu(2a)) + S(Cu(1a))·S(Cu(2))] - J(2c)S(Cu(2))·S(Cu(2a))), and J(3a) = -87.9(2) cm(-1), J(3b) = -5(1) cm(-1) and J(3c) = +5(3) cm(-1) in 3 (H = -J(3a)[S(Cu(1))·S(Cu(2)) + S(Cu(1a))·S(Cu(2a))] - J(3b)[S(Cu(1))·S(Cu(2a)) + S(Cu(1a))·S(Cu(2))] - J(3c)S(Cu(2))·S(Cu(2a))). For 1a, a net ferromagnetic coupling is observed with J(1a) = +0.86(1) cm(-1) (H = -J S(A)·S(B) + S(A)·D· S(B) + βH (g(A)S(A) + g(B)S(B)). This is the first example of ferromagnetic coupling in pyrophosphate-complexes reported to date. A structure-function correlation study focusing on magnetic exchange across the observed diverse pyrophosphate-bridges is described with density functional theory (DFT) calculations included to support the stated observations.
Sigma-Aldrich
Sodium pyrophosphate decahydrate, ACS reagent, ≥99%
Sigma-Aldrich
Sodium pyrophosphate tetrabasic, ≥95%
Sigma-Aldrich
Sodium pyrophosphate tetrabasic decahydrate, BioXtra, 99.0-103.0%
Sigma-Aldrich
Sodium pyrophosphate tetrabasic decahydrate, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Sodium pyrophosphate dibasic, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Sodium pyrophosphate dibasic, practical grade