Merck
Physico-chemical characterization of a new salt of ibuprofen.
Journal of pharmaceutical and biomedical analysis (1990-01-01)
M S Suleiman, N M Najib, M A Hassan, M E Abdel-Hamid
ABSTRACT
A new salt of ibuprofen was prepared by reaction with t-butylamine; its formation was confirmed by IR and 1H-NMR spectroscopy. The salt was characterized by thermoanalytical, X-ray powder diffraction and solubility studies. The salt was found to be 1.5 times more soluble in water than was ibuprofen, with an enthalpy of solution of -8.84 kcal mol-1.
Sigma-Aldrich
tert-Butylamine, ≥99.5%
Sigma-Aldrich
tert-Butylamine hydrochloride, ≥98.0% (AT)
Sigma-Aldrich
tert-Butylamine, 98%