Merck
Stress response of Nidula niveo-tomentosa to UV-A light.
Mycologia (2008-10-07)
Daniela E Taupp, Manfred Nimtz, Ralf G Berger, Holger Zorn
ABSTRACT
Exposition to UV-A light stimulated the growth and synthesis of raspberry ketone in submerged cultures of the basidiomycete Nidula niveotomentosa. To investigate the fungus' response to UV-A light differentially expressed proteins were identified by means of 2D-electrophoresis. Light induced proteins were de novo sequenced by ESI-MS/MS spectrometry, and the encoding nucleotide sequences were cloned from cDNA or genomic DNA. The spectrum of UV-A light-induced proteins comprised several stress-related proteins including a catalase, heat-shock proteins, glutathione S-transferases and proteasomes. In addition, growth-related enzymes of the citric cycle were found to be up-regulated as a response to irradiation with UV-A.
Sigma-Aldrich
4-(4-Hydroxyphenyl)-2-butanone, 99%
Sigma-Aldrich
4-(4-Hydroxyphenyl)-2-butanone, ≥98%, FCC, FG
Sigma-Aldrich
4-(4-Hydroxyphenyl)-2-butanone, natural, ≥98%, FCC, FG