Merck
  • Identification of a methioninase inhibitor, myrsinoic acid B, from Myrsine seguinii Lév., and its inhibitory activities.
Identification of a methioninase inhibitor, myrsinoic acid B, from Myrsine seguinii Lév., and its inhibitory activities.
Bioscience, biotechnology, and biochemistry (2008-09-09)
Satomi Ito, Atsushi Narise, Susumu Shimura
ABSTRACT
A methioninase inhibitor from Myrsine seguinii was purified and identified as myrsinoic acid B. Its inhibitory activities as to crude methioninase from periodontal bacteria such as Fusobacterium nucleatum, Porphyromonas gingivalis, and Treponema denticola were determined. The IC50 values were 10.5, 82.4, and 30.3 microM respectively.
Sigma-Aldrich
2-Oxobutyric acid, ≥95%, FG
Sigma-Aldrich
Sodium 2-oxobutyrate, powder
Sigma-Aldrich
2-Ketobutyric acid, 97%