Merck
  • Spectrophotometric determination of anilines based on charge-transfer reaction.
Spectrophotometric determination of anilines based on charge-transfer reaction.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2007-02-03)
Hao Wu, Li Ming Du
ABSTRACT
The molecular interactions between aniline, p-toluidines, benzidine and p-phenylenediamine as electron donors and 7,7,8,8-tetracyanoquinodimethane (TCNQ) as acceptor have been investigated by spectrophotometric method. Different variables affecting the reaction were studies and optimized. At the optimum reaction conditions Beer's law was obeyed in a concentration limit of 0.6-3.0, 0.3-3.0, 0.3-3.0 and 0.3-2.7 microg ml(-1) for aniline, p-toluidines, benzidine and p-phenylenediamine. The developed methods were applied successfully for the determination of the studied compounds in waste water and relative standard deviation of the methods were 0.8-3.0%. Percentage recoveries ranged from 97.22% to 102.78%.
Product Description
Supelco
p-Toluidine, for spectrophotometric det. of Au, Tl(III), W, ≥99.0%
Sigma-Aldrich
p-Toluidine hydrochloride, purum, ≥99.0% (AT)
Sigma-Aldrich
p-Toluidine, 99%
Sigma-Aldrich
p-Toluidine, 99.6%