Merck
  • Detection of mono- and di-hexoses as metabolites of 4-bromoaniline using HPLC-TOF-MS/MS.
Detection of mono- and di-hexoses as metabolites of 4-bromoaniline using HPLC-TOF-MS/MS.
Xenobiotica; the fate of foreign compounds in biological systems (2003-08-26)
H Major, J Castro-Perez, J K Nicholson, I D Wilson
ABSTRACT
1. The metabolic fate of 4-bromoaniline (4-BrA) was investigated in rat following intraperitoneal administration at 50 mg kg(-1) using HPLC-TOF-MS/MS. 2. The sensitivity provided by the use of TOF-MS/MS, aided by the distinctive isotope pattern resulting from the presence of the bromine substituent in the molecule, enabled the detection of many previously uncharacterized metabolites in the samples. 3. Several groups of minor metabolites were detected in the urine that corresponded to a number of isomeric hexose and di-hexose-containing conjugates (possibly glucosides and diglucosides) of 4-BrA. 4. As well as hexose and di-hexose conjugates of 4-BrA, several further groups of metabolites that also contained either a sulphamate or sulphate group in addition to the sugar moieties were also detected.
Sigma-Aldrich
4-Bromoaniline, 97%
Sigma-Aldrich
4-Bromoaniline, ≥99.0% (GC)