- Convenient assay for interferons.
Convenient assay for interferons.
Journal of virology (1981-02-01)
S Rubinstein, P C Familletti, S Pestka
PMID6163873
ABSTRACT
A convenient assay for interferons based on reduction of cytopathic effect was developed. The number of manipulations and the lengths of the various incubation steps were reduced to a minimum. The assay is simple to perform and can be completed within 16 h. Moreover, it can be used with various types of cells and a variety of viruses.
Sigma-Aldrich
Interferon αA from mouse, ≥95%, recombinant, expressed in E. coli, buffered aqueous solution, suitable for cell culture
Sigma-Aldrich
Interferon α from rat, ≥95%, recombinant, expressed in bacteria, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Interferon-αA human, ≥95%, suitable for cell culture, buffered aqueous solution, recombinant, expressed in E. coli
Sigma-Aldrich
Interferon-αA/D human, ≥95%, recombinant, expressed in E. coli, buffered aqueous solution, suitable for cell culture
Sigma-Aldrich
Interferon β from mouse, ≥95%, recombinant, expressed in E. coli, buffered aqueous solution, suitable for cell culture
Sigma-Aldrich
Interferon β from rat, ≥95%, recombinant, expressed in CHO cells, lyophilized powder, suitable for cell culture