Merck
  • Actomyosin contractility in olfactory placode neurons opens the skin epithelium to form the zebrafish nostril.
Actomyosin contractility in olfactory placode neurons opens the skin epithelium to form the zebrafish nostril.
Developmental cell (2023-02-26)
Marion Baraban, Clara Gordillo Pi, Isabelle Bonnet, Jean-François Gilles, Camille Lejeune, Mélody Cabrera, Florian Tep, Marie Anne Breau
ABSTRACT
Despite their barrier function, epithelia can locally lose their integrity to create physiological openings during morphogenesis. The mechanisms driving the formation of these epithelial breaks are only starting to be investigated. Here, we study the formation of the zebrafish nostril (the olfactory orifice), which opens in the skin epithelium to expose the olfactory neurons to external odorant cues. Combining live imaging, drug treatments, laser ablation, and tissue-specific functional perturbations, we characterize a mechanical interplay between olfactory placode neurons and the skin, which plays a crucial role in the formation of the orifice: the neurons pull on the overlying skin cells in an actomyosin-dependent manner which, in combination with a local reorganization of the skin epithelium, triggers the opening of the orifice. This work identifies an original mechanism to break an epithelial sheet, in which an adjacent group of cells mechanically assists the epithelium to induce its local rupture.
Sigma-Aldrich
(−)-Blebbistatin, solid, synthetic
Sigma-Aldrich
Anti-Laminin antibody produced in rabbit, 0.5 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-β-Catenin antibody produced in mouse, clone 15B8, ascites fluid
Sigma-Aldrich
Rho Kinase Inhibitor III, Rockout, The Rho Kinase Inhibitor III, Rockout, also referenced under CAS 7272-84-6, controls the biological activity of Rho Kinase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.
Sigma-Aldrich
Anti-phospho-Histone H3 (Ser10) Antibody, Mitosis Marker, Upstate®, from rabbit
Sigma-Aldrich
Anti-tp63 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
(Z)-4-Hydroxytamoxifen, ≥98% Z isomer