Merck
  • KIF11 UFMylation Maintains Photoreceptor Cilium Integrity and Retinal Homeostasis.
KIF11 UFMylation Maintains Photoreceptor Cilium Integrity and Retinal Homeostasis.
Advanced science (Weinheim, Baden-Wurttemberg, Germany) (2024-04-26)
Jie Ran, Guizhi Guo, Sai Zhang, Yufei Zhang, Liang Zhang, Dengwen Li, Shian Wu, Yusheng Cong, Xiaohong Wang, Songbo Xie, Huijie Zhao, Hongbin Liu, Guangshuo Ou, Xueliang Zhu, Jun Zhou, Min Liu
ABSTRACT
The photoreceptor cilium is vital for maintaining the structure and function of the retina. However, the molecular mechanisms underlying the photoreceptor cilium integrity and retinal homeostasis are largely unknown. Herein, it is shown that kinesin family member 11 (KIF11) localizes at the transition zone (connecting cilium) of the photoreceptor and plays a crucial role in orchestrating the cilium integrity. KIF11 depletion causes malformations of both the photoreceptor ciliary axoneme and membranous discs, resulting in photoreceptor degeneration and the accumulation of drusen-like deposits throughout the retina. Mechanistic studies show that the stability of KIF11 is regulated by an interplay between its UFMylation and ubiquitination; UFMylation of KIF11 at lysine 953 inhibits its ubiquitination by synoviolin 1 and thereby prevents its proteasomal degradation. The lysine 953-to-arginine mutant of KIF11 is more stable than wild-type KIF11 and also more effective in reversing the ciliary and retinal defects induced by KIF11 depletion. These findings identify a critical role for KIF11 UFMylation in the maintenance of photoreceptor cilium integrity and retinal homeostasis.
Sigma-Aldrich
Anti-UFL1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab2
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid