Merck
  • A role for mutations in AK9 and other genes affecting ependymal cells in idiopathic normal pressure hydrocephalus.
A role for mutations in AK9 and other genes affecting ependymal cells in idiopathic normal pressure hydrocephalus.
Proceedings of the National Academy of Sciences of the United States of America (2023-12-15)
Hong Wei Yang, Semin Lee, Bethany C Berry, Dejun Yang, Shaokuan Zheng, Rona S Carroll, Peter J Park, Mark D Johnson
ABSTRACT
Idiopathic normal pressure hydrocephalus (iNPH) is an enigmatic neurological disorder that develops after age 60 and is characterized by gait difficulty, dementia, and incontinence. Recently, we reported that heterozygous CWH43 deletions may cause iNPH. Here, we identify mutations affecting nine additional genes (AK9, RXFP2, PRKD1, HAVCR1, OTOG, MYO7A, NOTCH1, SPG11, and MYH13) that are statistically enriched among iNPH patients. The encoded proteins are all highly expressed in choroid plexus and ependymal cells, and most have been associated with cilia. Damaging mutations in AK9, which encodes an adenylate kinase, were detected in 9.6% of iNPH patients. Mice homozygous for an iNPH-associated AK9 mutation displayed normal cilia structure and number, but decreased cilia motility and beat frequency, communicating hydrocephalus, and balance impairment. AK9+/- mice displayed normal brain development and behavior until early adulthood, but subsequently developed communicating hydrocephalus. Together, our findings suggest that heterozygous mutations that impair ventricular epithelial function may contribute to iNPH.
Sigma-Aldrich
Anti-CWH43 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-AK9 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-SPG11 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution