Merck
  • Cognitive Impairment Is Associated with AMPAR Glutamatergic Dysfunction in a Mouse Model of Neuronal Methionine Synthase Deficiency.
Cognitive Impairment Is Associated with AMPAR Glutamatergic Dysfunction in a Mouse Model of Neuronal Methionine Synthase Deficiency.
Cells (2023-05-13)
Ziad Hassan, David Coelho, Carine Bossenmeyer-Pourié, Karim Matmat, Carole Arnold, Aurélie Savladori, Jean-Marc Alberto, Rémy Umoret, Jean-Louis Guéant, Grégory Pourié
ABSTRACT
Impairment of one-carbon metabolism during pregnancy, either due to nutritional deficiencies in B9 or B12 vitamins or caused by specific genetic defects, is often associated with neurological defects, including cognitive dysfunction that persists even after vitamin supplementation. Animal nutritional models do not allow for conclusions regarding the specific brain mechanisms that may be modulated by systemic compensations. Using the Cre-lox system associated to the neuronal promoter Thy1.2, a knock-out model for the methionine synthase specifically in the brain was generated. Our results on the neurobehavioral development of offspring show that the absence of methionine synthase did not lead to growth retardation, despite an effective reduction of both its expression and the methylation status in brain tissues. Behaviors were differently affected according to their functional outcome. Only temporary retardations were recorded in the acquisition of vegetative functions during the suckling period, compared to a dramatic reduction in cognitive performance after weaning. Investigation of the glutamatergic synapses in cognitive areas showed a reduction of AMPA receptors phosphorylation and clustering, indicating an epigenomic effect of the neuronal deficiency of methionine synthase on the reduction of glutamatergic synapses excitability. Altogether, our data indicate that cognitive impairment associated with methionine synthase deficiency may not only result from neurodevelopmental abnormalities, but may also be the consequence of alterations in functional plasticity of the brain.
Sigma-Aldrich
Anti-NMDAR2C Antibody, also recognizes NMDAR2A and 2B, Chemicon®, from rabbit
Sigma-Aldrich
Anti-NMDAR2B Antibody, phosphoTyr 1472, Chemicon®, from rabbit
Sigma-Aldrich
Anti-phospho NR1 (Ser897) Antibody, from rabbit, purified by affinity chromatography