Merck
  • Somatic mutations of CADM1 in aldosterone-producing adenomas and gap junction-dependent regulation of aldosterone production.
Somatic mutations of CADM1 in aldosterone-producing adenomas and gap junction-dependent regulation of aldosterone production.
Nature genetics (2023-06-09)
Xilin Wu, Elena A B Azizan, Emily Goodchild, Sumedha Garg, Man Hagiyama, Claudia P Cabrera, Fabio L Fernandes-Rosa, Sheerazed Boulkroun, Jyn Ling Kuan, Zenia Tiang, Alessia David, Masanori Murakami, Charles A Mein, Eva Wozniak, Wanfeng Zhao, Alison Marker, Folma Buss, Rebecca S Saleeb, Jackie Salsbury, Yuta Tezuka, Fumitoshi Satoh, Kenji Oki, Aaron M Udager, Debbie L Cohen, Heather Wachtel, Peter J King, William M Drake, Mark Gurnell, Jiri Ceral, Ales Ryska, Muaatamarulain Mustangin, Yin Ping Wong, Geok Chin Tan, Miroslav Solar, Martin Reincke, William E Rainey, Roger S Foo, Yutaka Takaoka, Sandra A Murray, Maria-Christina Zennaro, Felix Beuschlein, Akihiko Ito, Morris J Brown
ABSTRACT
Aldosterone-producing adenomas (APAs) are the commonest curable cause of hypertension. Most have gain-of-function somatic mutations of ion channels or transporters. Herein we report the discovery, replication and phenotype of mutations in the neuronal cell adhesion gene CADM1. Independent whole exome sequencing of 40 and 81 APAs found intramembranous p.Val380Asp or p.Gly379Asp variants in two patients whose hypertension and periodic primary aldosteronism were cured by adrenalectomy. Replication identified two more APAs with each variant (total, n = 6). The most upregulated gene (10- to 25-fold) in human adrenocortical H295R cells transduced with the mutations (compared to wildtype) was CYP11B2 (aldosterone synthase), and biological rhythms were the most differentially expressed process. CADM1 knockdown or mutation inhibited gap junction (GJ)-permeable dye transfer. GJ blockade by Gap27 increased CYP11B2 similarly to CADM1 mutation. Human adrenal zona glomerulosa (ZG) expression of GJA1 (the main GJ protein) was patchy, and annular GJs (sequelae of GJ communication) were less prominent in CYP11B2-positive micronodules than adjacent ZG. Somatic mutations of CADM1 cause reversible hypertension and reveal a role for GJ communication in suppressing physiological aldosterone production.
Sigma-Aldrich
Anti-GAPDH antibody, Mouse monoclonal, clone GAPDH-71.1, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody
Sigma-Aldrich
Anti-Mouse IgG (Fab specific)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Collagenase from Clostridium histolyticum, lyophilized powder (from 0.2 μm filtered solution), suitable for cell culture
Sigma-Aldrich
Anti-Connexin-43 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-SynCAM antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-GJA1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution