Merck
  • Notch Signaling Suppresses Melanoma Tumor Development in BRAF/Pten Mice.
Notch Signaling Suppresses Melanoma Tumor Development in BRAF/Pten Mice.
Cancers (2023-01-22)
Dareen Mikheil, Kirthana Prabhakar, Tun Lee Ng, Sireesh Teertam, B Jack Longley, Michael A Newton, Vijayasaradhi Setaluri
ABSTRACT
Both oncogenic and tumor suppressor roles have been assigned to Notch signaling in melanoma. In clinical trials, Notch inhibitors proved to be ineffective for melanoma treatment. Notch signaling has also been implicated in melanoma transdifferentiation, a prognostic feature in primary melanoma. In this study, we investigated the role of Notch signaling in melanoma tumor development and growth using the genetic model of mouse melanoma by crossing BRAFCA/+/Pten+/+/Tyr-CreER+ (B) and BRAFCA/+/Pten-/-/Tyr-CreER + (BP) mice with Notch1 or Notch2 floxed allele mice. The topical application of tamoxifen induced tumors in BP mice but not in B mice with or without the deletion of either Notch1 or Notch2. These data show that the loss of either Notch1 nor Notch2 can substitute the tumor suppressor function of Pten in BRAFV600E-induced melanomagenesis. However, in Pten-null background, the loss of either Notch1 or Notch2 appeared to accelerate BRAFV600E-induced tumor development, suggesting a tumor suppressor role for Notch1 and Notch2 in BRAFV600E/Pten-null driven melanomagenesis. Quantitative immunochemical analysis of a human cutaneous melanoma tissue microarray that consists of >100 primary tumors with complete clinical history showed a weak to moderate correlation between NOTCH protein levels and clinical and pathological parameters. Our data show that Notch signaling is involved during melanomagenesis and suggest that the identification of genes and signaling pathways downstream of Notch could help devise strategies for melanoma prevention.
Sigma-Aldrich
Anti-Notch 1 Antibody, NT, serum, from rabbit
Sigma-Aldrich
Anti-Notch1 Antibody, Intracellular, serum, from rabbit
Sigma-Aldrich
Anti-Notch 2 Antibody, NT, serum, from rabbit
Sigma-Aldrich
Anti-NOTCH2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution