Merck
GluNs Detection and Functions in Microglial Cells.
Methods in molecular biology (Clifton, N.J.) (2017-10-08)
Stéphane Peineau, Vincent Degos, Catherine Verney, Pierre Gressens
ABSTRACT
Proving endogenous GluN presence and functions in microglia require complementary steps to demonstrate (1) that GluN genes are transcripted and translated, (2) their cellular localization, (3) that the GluN are functional, and (4) the role of the functional GluN. The complete demonstration is performed by using mRNA detection technics, western blots, immunofluorescence, electrophysiology, calcium imaging, morphology studies, multiplex immunoassay together with conditional microglial Knock-Out mice and brain lesion models.
Sigma-Aldrich
Anti-NR3A Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-Glutamate Receptor NMDAR2A (NR2A) antibody produced in rabbit, affinity isolated antibody, lyophilized powder
Sigma-Aldrich
Anti-Glutamate Receptor NMDAR2C (NR2C) antibody produced in rabbit, affinity isolated antibody, lyophilized powder
Sigma-Aldrich
Anti-NR3B Antibody, Upstate®, from rabbit