Merck
  • Improved Protocol for Cardiac Differentiation and Maturation of Pluripotent Stem Cells.
Improved Protocol for Cardiac Differentiation and Maturation of Pluripotent Stem Cells.
Methods in molecular biology (Clifton, N.J.) (2019-05-28)
Imen Jebeniani, Shunli Ding, Michel Pucéat
ABSTRACT
Pluripotent stem cells feature the capacity to differentiate into any somatic cell types including cardiomyocytes. We report a cost-effective and simple protocol for the differentiation of specific ventricular cardiomyocytes. These cells are elongated, do not spontaneously beat, and do not feature any Ca2+-transient, an index of their stage of maturation toward adult cardiac cells. They represent a suitable model to screen both the efficiency and toxicology of drugs.
Sigma-Aldrich
Thiazovivin, ≥98% (HPLC)
Sigma-Aldrich
Fibronectin bovine plasma, solution, sterile-filtered, BioReagent, suitable for cell culture