Merck
  • Progesterone limits the tumor-promoting effects of the beta-subunit of human chorionic gonadotropin via non-nuclear receptors.
Progesterone limits the tumor-promoting effects of the beta-subunit of human chorionic gonadotropin via non-nuclear receptors.
iScience (2022-06-28)
Moumita Sarkar, Harsh Sharma, Parminder Singh, Ranbala Ranu, Ravi Datta Sharma, Usha Agrawal, Rahul Pal
ABSTRACT
The post-menopausal state in women is associated with increased cancer incidence, the reasons for which remain obscure. Curiously, increased circulating levels of beta-hCG (human chorionic gonadotropin) (a hormonal subunit linked with tumors of several lineages) are also often observed post-menopause. This study describes a previously unidentified interplay between beta-hCG and progesterone in tumorigenesis. Progesterone mediated apoptosis in beta-hCG responsive tumor cells via non-nuclear receptors. The transgenic expression of beta-hCG, particularly in the absence of the ovaries (a mimic of the post-menopausal state) constituted a potent pro-tumorigenic signal. Significantly, the administration of progesterone had significant anti-tumor effects. RNA-seq profiling identified molecular signatures associated with these processes. TCGA analysis revealed correlates between the expression of several newly identified genes and poor prognosis in post-menopausal patients of lung adenocarcinoma, colon adenocarcinoma, and glioblastoma. Specifically in these women, the detection of intra-tumoral/extra-tumoral beta-hCG may serve as a useful prognostic indicator, and treatment with progesterone on its detection may prove beneficial.
Sigma-Aldrich
Chorionic gonadotropin human, lyophilized powder, from human pregnancy urine
Sigma-Aldrich
JNK Inhibitor II, JNK Inhibitor II. SP600125, CAS 129-56-6, is a potent, cell-permeable, selective, and ATP competitive inhibitor of c-Jun N-terminal kinase (JNK; IC₅₀ = 40 nM for JNK-1 & JNK-2 & 90 nM for JNK-3).
Sigma-Aldrich
PD 98059, PD 98059, CAS 167869-21-8, is a cell-permeable, selective & reversible inhibitor of MAP Kinase Kinase (MEK). Inhibits MAP Kinase activation and subsequent phosphorylation of MAP Kinase substrates.
Sigma-Aldrich
SB 203580, SB 203580, CAS 152121-47-6, is a highly specific, potent, cell-permeable, selective, reversible, and ATP-competitive inhibitor of p38 MAP kinase (IC₅₀ = 34 nM in vitro, 600 nM in cells).
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 7, ≥98%
Sigma-Aldrich
Ro-20-1724, A cell-permeable, selective inhibitor of cAMP-specific phosphodiesterase (PDE IV; IC₅₀ = 2 µM).
Sigma-Aldrich
Caspase Inhibitor I, Z-VAD-FMK, CAS 187389-52-2, is a cell-permeable, irreversible, pan-caspase inhibitor.
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, A cell-permeable, non-specific inhibitor of cAMP and cGMP phosphodiesterases (IC₅₀ = 2-50 µM).