Merck
  • Palmitic acid conjugation enhances potency of tricyclo-DNA splice switching oligonucleotides.
Palmitic acid conjugation enhances potency of tricyclo-DNA splice switching oligonucleotides.
Nucleic acids research (2021-12-12)
Karima Relizani, Lucía Echevarría, Faouzi Zarrouki, Cécile Gastaldi, Chloe Dambrune, Philippine Aupy, Adrian Haeberli, Marek Komisarski, Thomas Tensorer, Thibaut Larcher, Fedor Svinartchouk, Cyrille Vaillend, Luis Garcia, Aurélie Goyenvalle
ABSTRACT
Tricyclo-DNA (tcDNA) is a conformationally constrained oligonucleotide analog that has demonstrated great therapeutic potential as antisense oligonucleotide (ASO) for several diseases. Like most ASOs in clinical development, tcDNA were modified with phosphorothioate (PS) backbone for therapeutic purposes in order to improve their biodistribution by enhancing association with plasma and cell protein. Despite the advantageous protein binding properties, systemic delivery of PS-ASO remains limited and PS modifications can result in dose limiting toxicities in the clinic. Improving extra-hepatic delivery of ASO is highly desirable for the treatment of a variety of diseases including neuromuscular disorders such as Duchenne muscular dystrophy. We hypothesized that conjugation of palmitic acid to tcDNA could facilitate the delivery of the ASO from the bloodstream to the interstitium of the muscle tissues. We demonstrate here that palmitic acid conjugation enhances the potency of tcDNA-ASO in skeletal and cardiac muscles, leading to functional improvement in dystrophic mice with significantly reduced dose of administered ASO. Interestingly, palmitic acid-conjugated tcDNA with a full phosphodiester backbone proved effective with a particularly encouraging safety profile, offering new perspectives for the clinical development of PS-free tcDNA-ASO for neuromuscular diseases.
Product Description
Millipore
MILLIPLEX® Mouse Kidney Injury Magnetic Bead Panel 1 - Toxicity Multiplex Assay, The analytes available for this multiplex kit are: β-2-Microglobulin, IP-10, KIM-1, Renin, TIMP-1, VEGF (for urine samples) or IP-10, KIM-1, Renin, and TIMP-1 (for serum/plasma samples).
Millipore
MILLIPLEX® Mouse Kidney Injury Magnetic Bead Panel 2 - Toxicity Multiplex Assay, The analytes available for this multiplex kit are: Clusterin, Cystatin C, EGF, Lipocalin-2/NGAL, Osteopontin (OPN) (for urine samples) or Clusterin, Cystatin C, Lipocalin-2/NGAL, Osteopontin (OPN) (for serum/plasma samples).