Merck
Endotoxins and disease mechanisms.
Annual review of medicine (1987-01-01)
D C Morrison, J L Ryan
ABSTRACT
In this chapter, current concepts about the mechanisms of action of endotoxin are reviewed. Particular attention is focused upon endotoxin-induced production of soluble mediators from macrophages and mononuclear cells and on the potential contribution of these mediators to endotoxin shock. In many cases, the interrelationships between these mediators as primary and/or secondary consequences of endotoxin stimulation of mononuclear phagocytes are discussed. Final comments address the relevance of these mediators to the therapy of endotoxin shock.
Sigma-Aldrich
Proteus FISH probe - ATTO488, Probe for fluorescence in situ hybridization (FISH),20 μM in water
Sigma-Aldrich
Microbial DNA standard from Proteus vulgaris, Suitable for PCR, sequencing and NGS
Sigma-Aldrich
Lipopolysaccharides from Proteus vulgaris, purified by phenol extraction
Sigma-Aldrich
Microbial DNA standard from Proteus mirabilis, Suitable for PCR, sequencing and NGS
Sigma-Aldrich
Proteus FISH probe - Cy3, Probe for fluorescence in situ hybridization (FISH),20 μM in water
Sigma-Aldrich
Lipopolysaccharides from Proteus mirabilis, purified by phenol extraction