Merck
  • Extracellular domain shedding of the ALK receptor mediates neuroblastoma cell migration.
Extracellular domain shedding of the ALK receptor mediates neuroblastoma cell migration.
Cell reports (2021-07-15)
Hao Huang, Alexander Gont, Lynn Kee, Ruben Dries, Kathrin Pfeifer, Bandana Sharma, David N Debruyne, Matthew Harlow, Satyaki Sengupta, Jikui Guan, Caleb M Yeung, Wenchao Wang, Bengt Hallberg, Ruth H Palmer, Meredith S Irwin, Rani E George
ABSTRACT
Although activating mutations of the anaplastic lymphoma kinase (ALK) membrane receptor occur in ∼10% of neuroblastoma (NB) tumors, the role of the wild-type (WT) receptor, which is aberrantly expressed in most non-mutated cases, is unclear. Both WT and mutant proteins undergo extracellular domain (ECD) cleavage. Here, we map the cleavage site to Asn654-Leu655 and demonstrate that cleavage inhibition of WT ALK significantly impedes NB cell migration with subsequent prolongation of survival in mouse models. Cleavage inhibition results in the downregulation of an epithelial-to-mesenchymal transition (EMT) gene signature, with decreased nuclear localization and occupancy of β-catenin at EMT gene promoters. We further show that cleavage is mediated by matrix metalloproteinase 9, whose genetic and pharmacologic inactivation inhibits cleavage and decreases NB cell migration. Together, our results indicate a pivotal role for WT ALK ECD cleavage in NB pathogenesis, which may be harnessed for therapeutic benefit.
Product Description
Millipore
FLAG® M Purification Kit, For Mammalian expression systems.
Sigma-Aldrich
MMP-12 Inhibitor, MMP408, The MMP-12 Inhibitor, MMP408, also referenced under CAS 1258003-93-8, controls the biological activity of MMP-12. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.
Sigma-Aldrich
Decamethylcyclopentasiloxane, 97%
Millipore
MMP-9/MMP-13 Inhibitor I, The MMP-9/MMP-13 Inhibitor I, also referenced under CAS 204140-01-2, controls the biological activity of MMP-9/MMP-13. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.
Sigma-Aldrich
QCM ECMatrix Cell Invasion Assay, 24-well (8 µm), fluorimetric, The Cell Invasion Assay Kit kuses a 24 well plate with 8 um pores, which is ideal for evaluation of invasive tumor cells.