Merck
  • The translocon-associated protein (TRAP) complex regulates quality control of N-linked glycosylation during ER stress.
The translocon-associated protein (TRAP) complex regulates quality control of N-linked glycosylation during ER stress.
Science advances (2021-02-02)
Chatchai Phoomak, Wei Cui, Thomas J Hayman, Seok-Ho Yu, Peng Zhao, Lance Wells, Richard Steet, Joseph N Contessa
ABSTRACT
Asparagine (N)-linked glycosylation is required for endoplasmic reticulum (ER) homeostasis, but how this co- and posttranslational modification is maintained during ER stress is unknown. Here, we introduce a fluorescence-based strategy to detect aberrant N-glycosylation in individual cells and identify a regulatory role for the heterotetrameric translocon-associated protein (TRAP) complex. Unexpectedly, cells with knockout of SSR3 or SSR4 subunits restore N-glycosylation over time concurrent with a diminished ER stress transcriptional signature. Activation of ER stress or silencing of the ER chaperone BiP exacerbates or rescues the glycosylation defects, respectively, indicating that SSR3 and SSR4 enable N-glycosylation during ER stress. Protein levels of the SSR3 subunit are ER stress and UBE2J1 dependent, revealing a mechanism that coordinates upstream N-glycosylation proficiency with downstream ER-associated degradation and proteostasis. The fidelity of N-glycosylation is not static in both nontransformed and tumor cells, and the TRAP complex regulates ER glycoprotein quality control under conditions of stress.
Sigma-Aldrich
MISSION® esiRNA, targeting human HSPA5
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
Duolink® PLA Multicolor Reagent Pack
Sigma-Aldrich
Anti-SSR3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Duolink® In Situ Mounting Medium with DAPI
Sigma-Aldrich
Duolink® PLA Multicolor Probemaker Kit - Red, Proximity Ligation Assay Multiplexing: protein-protein interaction, post-translational modification and low abundant protein events
Sigma-Aldrich
Thapsigargin, ≥98% (HPLC), solid film