Merck
  • Polystyrene-Based Hydroxide-Ion-Conducting Ionomer: Binder Characteristics and Performance in Anion-Exchange Membrane Fuel Cells.
Polystyrene-Based Hydroxide-Ion-Conducting Ionomer: Binder Characteristics and Performance in Anion-Exchange Membrane Fuel Cells.
Polymers (2021-03-07)
Ji Eon Chae, So Young Lee, Sung Jong Yoo, Jin Young Kim, Jong Hyun Jang, Hee-Young Park, Hyun Seo Park, Bora Seo, Dirk Henkensmeier, Kwang Ho Song, Hyoung-Juhn Kim
ABSTRACT
Polystyrene-based polymers with variable molecular weights are prepared by radical polymerization of styrene. Polystyrene is grafted with bromo-alkyl chains of different lengths through Friedel-Crafts acylation and quaternized to afford a series of hydroxide-ion-conducting ionomers for the catalyst binder for the membrane electrode assembly in anion-exchange membrane fuel cells (AEMFCs). Structural analyses reveal that the molecular weight of the polystyrene backbone ranges from 10,000 to 63,000 g mol-1, while the ion exchange capacity of quaternary-ammonium-group-bearing ionomers ranges from 1.44 to 1.74 mmol g-1. The performance of AEMFCs constructed using the prepared electrode ionomers is affected by several ionomer properties, and a maximal power density of 407 mW cm-2 and a durability exceeding that of a reference cell with a commercially available ionomer are achieved under optimal conditions. Thus, the developed approach is concluded to be well suited for the fabrication of next-generation electrode ionomers for high-performance AEMFCs.
Sigma-Aldrich
3-Bromopropionyl chloride, technical grade
Sigma-Aldrich
Benzenesulfonyl chloride, 99%
Sigma-Aldrich
Aluminum chloride, ReagentPlus®, 99%
Sigma-Aldrich
D.E.R. 332, used as embedding medium
Sigma-Aldrich
6-Bromohexanoyl chloride, 97%