Merck
  • Postnatal expression and possible function of RANK and RANKL in the murine inner ear.
Postnatal expression and possible function of RANK and RANKL in the murine inner ear.
Bone (2021-01-02)
Shyan-Yuan Kao, Sachiyo Katsumi, Dongjun Han, Argyro J Bizaki-Vallaskangas, Sasa Vasilijic, Lukas D Landegger, Arthur G Kristiansen, Michael J McKenna, Konstantina M Stankovic
ABSTRACT
The bone encasing the inner ear, known as the otic capsule, is unique because it remodels little postnatally compared to other bones in the body. Previous studies established that osteoprotegerin (OPG) in the inner ear inhibits otic capsule remodeling. OPG acts as a decoy receptor of receptor activator of nuclear factor κB ligand (RANKL) to disrupt the interaction between RANKL and RANK, the primary regulators of bone metabolism. Here we studied the expression and function of RANK and RANKL in the murine cochlea. Using a combination of in situ hybridization, real-time quantitative RT-PCR, and western blot, we demonstrate that Rankl and Rank genes and their protein products are expressed in the intracochlear soft tissues and the otic capsule in a developmentally regulated manner. Using a culture of neonatal murine cochlear neurons, we show that the interaction between RANK and RANKL inhibits neurite outgrowth in these neurons, and is associated with upregulation of NOGO-A expression. Taken together, our results suggest that, in addition to regulating otic capsule bone remodeling, RANK and RANKL expressed by intracochlear soft tissues may also regulate spiral ganglion neuron function by affecting neurite outgrowth.
Sigma-Aldrich
Brain-derived neurotrophic factor human, BDNF, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
NT-3 from mouse, recombinant, expressed in E. coli, ≥97% (SDS-PAGE), ≥97% (HPLC)
Sigma-Aldrich
Fast Red TR Salt 1,5-naphthalenedisulfonate salt
Sigma-Aldrich
Horse Serum, Donor herd, USA origin, sterile-filtered, suitable for cell culture, suitable for hybridoma
Sigma-Aldrich
5-Norbornene-2-endo,3-endo-dimethanol, 98%