Merck
  • Extracellular Matrix Injury of Kidney Allografts in Antibody-Mediated Rejection: A Proteomics Study.
Extracellular Matrix Injury of Kidney Allografts in Antibody-Mediated Rejection: A Proteomics Study.
Journal of the American Society of Nephrology : JASN (2020-09-10)
Sergi Clotet-Freixas, Caitriona M McEvoy, Ihor Batruch, Chiara Pastrello, Max Kotlyar, Julie Anh Dung Van, Madhurangi Arambewela, Alex Boshart, Sofia Farkona, Yun Niu, Yanhong Li, Olusegun Famure, Andrea Bozovic, Vathany Kulasingam, Peixuen Chen, S Joseph Kim, Emilie Chan, Sajad Moshkelgosha, Syed Ashiqur Rahman, Jishnu Das, Tereza Martinu, Stephen Juvet, Igor Jurisica, Andrzej Chruscinski, Rohan John, Ana Konvalinka
ABSTRACT
Antibody-mediated rejection (AMR) accounts for >50% of kidney allograft loss. Donor-specific antibodies (DSA) against HLA and non-HLA antigens in the glomeruli and the tubulointerstitium cause AMR while inflammatory cytokines such as TNFα trigger graft injury. The mechanisms governing cell-specific injury in AMR remain unclear. Unbiased proteomic analysis of laser-captured and microdissected glomeruli and tubulointerstitium was performed on 30 for-cause kidney biopsy specimens with early AMR, acute cellular rejection (ACR), or acute tubular necrosis (ATN). A total of 107 of 2026 glomerular and 112 of 2399 tubulointerstitial proteins was significantly differentially expressed in AMR versus ACR; 112 of 2026 glomerular and 181 of 2399 tubulointerstitial proteins were significantly dysregulated in AMR versus ATN (P<0.05). Basement membrane and extracellular matrix (ECM) proteins were significantly decreased in both AMR compartments. Glomerular and tubulointerstitial laminin subunit γ-1 (LAMC1) expression decreased in AMR, as did glomerular nephrin (NPHS1) and receptor-type tyrosine-phosphatase O (PTPRO). The proteomic analysis revealed upregulated galectin-1, which is an immunomodulatory protein linked to the ECM, in AMR glomeruli. Anti-HLA class I antibodies significantly increased cathepsin-V (CTSV) expression and galectin-1 expression and secretion in human glomerular endothelial cells. CTSV had been predicted to cleave ECM proteins in the AMR glomeruli. Glutathione S-transferase ω-1, an ECM-modifying enzyme, was significantly increased in the AMR tubulointerstitium and in TNFα-treated proximal tubular epithelial cells. Basement membranes are often remodeled in chronic AMR. Proteomic analysis performed on laser-captured and microdissected glomeruli and tubulointerstitium identified early ECM remodeling, which may represent a new therapeutic opportunity.
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-GSTO1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-GAPDH Mouse mAb (6C5), liquid, clone 6C5, Calbiochem®
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-LAMC1 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-GLEPP1/PTPRO Antibody, clone 5C11, clone 5C11, from mouse