Merck
  • Intranasal mesenchymal stem cell therapy to boost myelination after encephalopathy of prematurity.
Intranasal mesenchymal stem cell therapy to boost myelination after encephalopathy of prematurity.
Glia (2020-10-13)
Josine E G Vaes, Caren M van Kammen, Chloe Trayford, Annette van der Toorn, Torben Ruhwedel, Manon J N L Benders, Rick M Dijkhuizen, Wiebke Möbius, Sabine H van Rijt, Cora H Nijboer
ABSTRACT
Encephalopathy of prematurity (EoP) is a common cause of long-term neurodevelopmental morbidity in extreme preterm infants. Diffuse white matter injury (dWMI) is currently the most commonly observed form of EoP. Impaired maturation of oligodendrocytes (OLs) is the main underlying pathophysiological mechanism. No therapies are currently available to combat dWMI. Intranasal application of mesenchymal stem cells (MSCs) is a promising therapeutic option to boost neuroregeneration after injury. Here, we developed a double-hit dWMI mouse model and investigated the therapeutic potential of intranasal MSC therapy. Postnatal systemic inflammation and hypoxia-ischemia led to transient deficits in cortical myelination and OL maturation, functional deficits and neuroinflammation. Intranasal MSCs migrated dispersedly into the injured brain and potently improved myelination and functional outcome, dampened cerebral inflammationand rescued OL maturation after dWMI. Cocultures of MSCs with primary microglia or OLs show that MSCs secrete factors that directly promote OL maturation and dampen neuroinflammation. We show that MSCs adapt their secretome after ex vivo exposure to dWMI milieu and identified several factors including IGF1, EGF, LIF, and IL11 that potently boost OL maturation. Additionally, we showed that MSC-treated dWMI brains express different levels of these beneficial secreted factors. In conclusion, the combination of postnatal systemic inflammation and hypoxia-ischemia leads to a pattern of developmental brain abnormalities that mimics the clinical situation. Intranasal delivery of MSCs, that secrete several beneficial factors in situ, is a promising strategy to restore myelination after dWMI and subsequently improve the neurodevelopmental outcome of extreme preterm infants in the future.
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)
Sigma-Aldrich
Trypsin from bovine pancreas, TPCK Treated, essentially salt-free, lyophilized powder, ≥10,000 BAEE units/mg protein
Sigma-Aldrich
Monoclonal Anti-MAP2 antibody produced in mouse, clone HM-2, ascites fluid
Sigma-Aldrich
Paraformaldehyde, meets analytical specification of DAC, 95.0-100.5%
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
N-Acetyl-L-cysteine, BioXtra, ≥99% (TLC)
Sigma-Aldrich
Poly-L-ornithine hydrobromide, mol wt 30,000-70,000
Sigma-Aldrich
Trypsin from porcine pancreas, lyophilized powder, BioReagent, suitable for cell culture, 1,000-2,000 BAEE units/mg solid
Sigma-Aldrich
Deoxyribonuclease I from bovine pancreas, Type IV, lyophilized powder, ≥2,000 Kunitz units/mg protein
Sigma-Aldrich
Poly-D-lysine hydrobromide, mol wt 70,000-150,000, lyophilized powder, γ-irradiated, BioReagent, suitable for cell culture
Sigma-Aldrich
Brij® L23, main component: tricosaethylene glycol dodecyl ether
Sigma-Aldrich
Poly-DL-ornithine hydrobromide, mol wt 15,000-30,000
Sigma-Aldrich
Anti-Myelin Basic Protein Antibody, a.a. 82-87, culture supernatant, clone 12, Chemicon®
Sigma-Aldrich
Anti-Olig-2 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Olig2 Antibody, clone 211F1.1, clone 211F1.1, from mouse