Merck
Maternal obesity impairs skeletal development in adult offspring
The Journal of endocrinology (2018-10-12)
Jin-Ran Chen, Oxana P Lazarenko, Haijun Zhao, Alexander W Alund, Kartik Shankar
ABSTRACT
Intrauterine or early postnatal high-fat diet (HFD) has substantial influences on adult offspring health; however, studies of HFD-induced maternal obesity on regulation of adult offspring bone formation are sparse. Here, we investigated the effects of HFD-induced maternal obesity on both fetal and adult offspring skeletal development. We found that HFD-induced maternal obesity significantly decreased fetal skeletal development, but enhanced fetal osteoblastic cell senescence signaling and significantly increased the expression of inflammatory factors of the senescence-associated secretory phenotype (SASP) in osteo-progenitors. It was found that p300/CBP activation led to H3K27 acetylation to increase the expression of senescence-related genes and PPARγ in embryonic mouse osteogenic calvarial cells from HFD obese dams. These results were recapitulated in human umbilical cord mesenchymal stem cells (UC MSCs) isolated from offspring of pregnant obese and lean mothers following delivery. Regardless of postnatal HFD challenge, adult offspring from HFD obese dams showed significantly suppressed bone formation. Such early involution of bone formation of adult offspring from HFD obese dams may at least in part due to histone acetylation, i.e., epigenetic regulation of genes involved in cell senescence signaling in pre-osteoblasts from prenatal development. These findings indicate fetal pre-osteoblastic cell senescence signaling is epigenetically regulated by maternal obesity to repress bone formation in adult offspring in rodents and suggest that at least some of these effects may also manifest in humans.
Sigma-Aldrich
Anti-CBP antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Osteocalcin Antibody, serum, from rabbit
Sigma-Aldrich
Anti-p300/CBP antibody, Mouse monoclonal, clone NM11, purified from hybridoma cell culture
Sigma-Aldrich
Anti-p300 CT Antibody, clone RW128, clone RW128, Upstate®, from mouse
Sigma-Aldrich
Anti-acetyl-Histone H3 (Lys27) Antibody, serum, Upstate®