Merck
  • A rapid and highly sensitive UPLC-ESI-MS/MS method for the analysis of the fatty acid profile of edible vegetable oils.
A rapid and highly sensitive UPLC-ESI-MS/MS method for the analysis of the fatty acid profile of edible vegetable oils.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2020-11-28)
Suresh K Nagumalli, Cristina C Jacob, Gonçalo Gamboa da Costa, Suresh K Nagumalli, Cristina C Jacob, Gonçalo Gamboa da Costa
ABSTRACT
The analysis of the fatty acid profile of triglycerides has long played a central role in the evaluation and classification of edible vegetable oils. However, the range of analytical procedures available to evaluate these profiles remains limited and are typically based on transesterification of the triglyceride fatty acid residues to methyl esters, followed by capillary gas-liquid chromatography (GC) coupled with flame ionization or mass spectrometry detection. Although robust and long-proven, these analytical methods tend to entail long chromatographic runs and are relatively insensitive. In order to expand the range of available techniques for the analysis of the fatty acid profile of triglycerides in vegetable oils, we report herein a novel method based upon a rapid and straightforward transesterification of the triglycerides with dimethylaminoethanol under alkaline conditions, followed by a "dilute-and-shoot" analysis by ultra-performance liquid chromatography coupled with electrospray tandem mass spectrometry. The chromatographic analysis is accomplished in 1.5 min, affording a high throughput of samples compared to techniques based upon GC approaches. The method performance was assessed intra- and inter-day with 10 representative saturated and unsaturated fatty acids ranging from C8 to C18 and afforded fatty acid profile accuracies of 93-108% and imprecisions of only 0.3-2.0%. The limit of quantification of the method, estimated as the minimum amount of derivatized oil sample capable of affording less than 20% accuracy and precision error was determined to be approximately 0.5 pg on-column, making this new method potentially valuable for fields where high sensitivity, precision, and accuracy may be required, such as in toxicology studies, forensics, archeology, or art analysis.
Sigma-Aldrich
Glyceryl tridodecanoate, ≥99%
Sigma-Aldrich
Glyceryl trimyristate, ≥99%
Sigma-Aldrich
Glyceryl tripalmitoleate, ≥98%, liquid
Sigma-Aldrich
Glyceryl trilinolenate, ≥97% (TLC), liquid