Merck
  • Glycolysis regulates Hedgehog signalling via the plasma membrane potential.
Glycolysis regulates Hedgehog signalling via the plasma membrane potential.
The EMBO journal (2020-10-07)
Stephanie Spannl, Tomasz Buhl, Ioannis Nellas, Salma A Zeidan, K Venkatesan Iyer, Helena Khaliullina, Carsten Schultz, André Nadler, Natalie A Dye, Suzanne Eaton
ABSTRACT
Changes in cell metabolism and plasma membrane potential have been linked to shifts between tissue growth and differentiation, and to developmental patterning. How such changes mediate these effects is poorly understood. Here, we use the developing wing of Drosophila to investigate the interplay between cell metabolism and a key developmental regulator-the Hedgehog (Hh) signalling pathway. We show that reducing glycolysis both lowers steady-state levels of ATP and stabilizes Smoothened (Smo), the 7-pass transmembrane protein that transduces the Hh signal. As a result, the transcription factor Cubitus interruptus accumulates in its full-length, transcription activating form. We show that glycolysis is required to maintain the plasma membrane potential and that plasma membrane depolarization blocks cellular uptake of N-acylethanolamides-lipoprotein-borne Hh pathway inhibitors required for Smo destabilization. Similarly, pharmacological inhibition of glycolysis in mammalian cells induces ciliary translocation of Smo-a key step in pathway activation-in the absence of Hh. Thus, changes in cell metabolism alter Hh signalling through their effects on plasma membrane potential.
Sigma-Aldrich
Bromopyruvic acid, ≥98.0%
Sigma-Aldrich
Gramicidin A from Bacillus brevis
Sigma-Aldrich
Anti-Acetylated Tubulin antibody, Mouse monoclonal, clone 6-11B-1, purified from hybridoma cell culture
Sigma-Aldrich
Donkey Anti-Goat IgG Antibody, HRP conjugate, Species Adsorbed, Chemicon®, from donkey
Sigma-Aldrich
Bis(1,3-dibutylbarbituric acid) trimethine oxonol, ≥95% (HPLC)
Sigma-Aldrich
Antimycin A from Streptomyces sp.
NAP-5 20 ST, Cytiva 17-0853-01