Merck
  • FcRn is a CD32a coreceptor that determines susceptibility to IgG immune complex-driven autoimmunity.
FcRn is a CD32a coreceptor that determines susceptibility to IgG immune complex-driven autoimmunity.
The Journal of experimental medicine (2020-07-14)
Jonathan J Hubbard, Michal Pyzik, Timo Rath, Lisa K Kozicky, Kine M K Sand, Amit K Gandhi, Algirdas Grevys, Stian Foss, Susan C Menzies, Jonathan N Glickman, Edda Fiebiger, Derry C Roopenian, Inger Sandlie, Jan Terje Andersen, Laura M Sly, Kristi Baker, Richard S Blumberg
ABSTRACT
IgG immune complexes (ICs) promote autoimmunity through binding fragment crystallizable (Fc) γ-receptors (FcγRs). Of these, the highly prevalent FcγRIIa (CD32a) histidine (H)-131 variant (CD32aH) is strongly linked to human autoimmune diseases through unclear mechanisms. We show that, relative to the CD32a arginine (R)-131 (CD32aR) variant, CD32aH more avidly bound human (h) IgG1 IC and formed a ternary complex with the neonatal Fc receptor (FcRn) under acidic conditions. In primary human and mouse cells, both CD32a variants required FcRn to induce innate and adaptive immune responses to hIgG1 ICs, which were augmented in the setting of CD32aH. Conversely, FcRn induced responses to IgG IC independently of classical FcγR, but optimal responses required FcRn and FcγR. Finally, FcRn blockade decreased inflammation in a rheumatoid arthritis model without reducing circulating autoantibody levels, providing support for FcRn's direct role in IgG IC-associated inflammation. Thus, CD32a and FcRn coregulate IgG IC-mediated immunity in a manner favoring the CD32aH variant, providing a novel mechanism for its disease association.
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-FCGRT antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available.
Sigma-Aldrich
Duolink® In Situ Detection Reagents FarRed
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-FCGR2A/C Antibody, clone 11B6, clone 11B6, from mouse
Pricing and availability is not currently available.