Merck
  • 3D imaging of colorectal cancer organoids identifies responses to Tankyrase inhibitors.
3D imaging of colorectal cancer organoids identifies responses to Tankyrase inhibitors.
PloS one (2020-08-19)
Luned M Badder, Andrew J Hollins, Bram Herpers, Kuan Yan, Kenneth B Ewan, Mairian Thomas, Jennifer R Shone, Delyth A Badder, Marc Naven, Kevin E Ashelford, Rachel Hargest, Alan R Clarke, Christina Esdar, Hans-Peter Buchstaller, J Mark Treherne, Sylvia Boj, Bahar Ramezanpour, Dirk Wienke, Leo S Price, Paul H Shaw, Trevor C Dale
ABSTRACT
Aberrant activation of the Wnt signalling pathway is required for tumour initiation and survival in the majority of colorectal cancers. The development of inhibitors of Wnt signalling has been the focus of multiple drug discovery programs targeting colorectal cancer and other malignancies associated with aberrant pathway activation. However, progression of new clinical entities targeting the Wnt pathway has been slow. One challenge lies with the limited predictive power of 2D cancer cell lines because they fail to fully recapitulate intratumoural phenotypic heterogeneity. In particular, the relationship between 2D cancer cell biology and cancer stem cell function is poorly understood. By contrast, 3D tumour organoids provide a platform in which complex cell-cell interactions can be studied. However, complex 3D models provide a challenging platform for the quantitative analysis of drug responses of therapies that have differential effects on tumour cell subpopulations. Here, we generated tumour organoids from colorectal cancer patients and tested their responses to inhibitors of Tankyrase (TNKSi) which are known to modulate Wnt signalling. Using compounds with 3 orders of magnitude difference in cellular mechanistic potency together with image-based assays, we demonstrate that morphometric analyses can capture subtle alterations in organoid responses to Wnt inhibitors that are consistent with activity against a cancer stem cell subpopulation. Overall our study highlights the value of phenotypic readouts as a quantitative method to asses drug-induced effects in a relevant preclinical model.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
3dGRO® Human CRC Organoids (ISO50)
Sigma-Aldrich
3dGRO® Human CRC Organoids (ISO34)
Sigma-Aldrich
3dGRO® Human CRC Organoids (ISO68)
Sigma-Aldrich
3dGRO® Human CRC Organoids (ISO38)
Sigma-Aldrich
3dGRO® Human CRC Organoids (ISO57)
Sigma-Aldrich
3dGRO® Human CRC Organoids (ISO72)
Sigma-Aldrich
3dGRO® Human CRC Organoids (ISO49)
Sigma-Aldrich
3dGRO® Human CRC Organoids (ISO48)