Merck
  • SMCHD1 promotes ATM-dependent DNA damage signaling and repair of uncapped telomeres.
SMCHD1 promotes ATM-dependent DNA damage signaling and repair of uncapped telomeres.
The EMBO journal (2020-02-23)
Aleksandra Vančevska, Wareed Ahmed, Verena Pfeiffer, Marianna Feretzaki, Simon J Boulton, Joachim Lingner
ABSTRACT
Structural maintenance of chromosomes flexible hinge domain-containing protein 1 (SMCHD1) has been implicated in X-chromosome inactivation, imprinting, and DNA damage repair, and mutations in SMCHD1 can cause facioscapulohumeral muscular dystrophy. More recently, SMCHD1 has also been identified as a component of telomeric chromatin. Here, we report that SMCHD1 is required for DNA damage signaling and non-homologous end joining (NHEJ) at unprotected telomeres. Co-depletion of SMCHD1 and the shelterin subunit TRF2 reduced telomeric 3'-overhang removal in time-course experiments, as well as the number of chromosome end fusions. SMCHD1-deficient cells displayed reduced ATM S1981 phosphorylation and diminished formation of γH2AX foci and of 53BP1-containing telomere dysfunction-induced foci (TIFs), indicating defects in DNA damage checkpoint signaling. Removal of TPP1 and subsequent activation of ATR signaling rescued telomere fusion events in TRF2-depleted SMCHD1 knockout cells. Together, these data indicate that SMCHD1 depletion reduces telomere fusions in TRF2-depleted cells due to defects in ATM-dependent checkpoint signaling and that SMCHD1 mediates DNA damage response activation upstream of ATM phosphorylation at uncapped telomeres.
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse
Sigma-Aldrich
Anti-TRF2 Antibody, clone 4A794, clone 4A794, Upstate®, from mouse
Sigma-Aldrich
Demecolcine, ≥98% (HPLC)
Sigma-Aldrich
Anti-Chk2 Antibody, clone 7, clone 7, Upstate®, from mouse