Merck
  • Extracellular vesicles from oviductal isthmus and ampulla stimulate the induced acrosome reaction and signaling events associated with capacitation in bovine spermatozoa.
Extracellular vesicles from oviductal isthmus and ampulla stimulate the induced acrosome reaction and signaling events associated with capacitation in bovine spermatozoa.
Journal of cellular biochemistry (2019-11-07)
A Franchi, A Moreno-Irusta, E M Domínguez, A J Adre, L C Giojalas
ABSTRACT
Cells can communicate with other neighboring or distant cells through the secretion of extracellular vesicles (EV), composed of a lipid bilayer and bearing surface molecules that allow them to recognize target cells. In this way, EV induce signaling via different mechanisms, modulating the physiological state of the recipient cell. EV have been identified in both male and female reproductive fluids, however, the possible role of EV isolated from female reproductive fluids has become an emerging field only recently. It is known that ejaculated mammalian spermatozoa need to undergo physiological preparation in the female reproductive tract to fertilize the egg. EV secreted by different regions of the female tract constitute signals that may have a key role in regulating sperm functions. The aims of the present study were isolating EV from different regions of the bovine oviduct and analyzing their interaction and physiological effects on spermatozoa. Here, we report the characterization of bovine oviductal fluid EV from the isthmus and ampulla region and their effect on the induced acrosome reaction and signaling events associated with sperm capacitation. EV induced an increase in sperm protein tyrosine phosphorylation, while cell survival of cryopreserved bovine spermatozoa was maintained. We also show that EV uptake regulates the sperm calcium levels by inducing an immediate increase in the intracellular calcium concentration and sperm priming, after a pre-incubation period, of the progesterone-induced intracellular calcium rise. Our data contribute to understand the role of EV in the communication between the female reproductive tract and the sperm physiology, information that may be used to improve the efficiency of reproductive assisted technologies.
Sigma-Aldrich
Luminol, ≥97% (HPLC)
Sigma-Aldrich
Anti-Heat Shock Protein 70 Antibody, clone W27, clone W27, Chemicon®, from mouse
Sigma-Aldrich
p-Coumaric acid, ≥98.0% (HPLC)
Sigma-Aldrich
Anti-Phosphotyrosine Antibody, clone 4G10®, clone 4G10®, Upstate®, from mouse