Merck
  • Conserved Lipid and Small-Molecule Modulation of COQ8 Reveals Regulation of the Ancient Kinase-like UbiB Family.
Conserved Lipid and Small-Molecule Modulation of COQ8 Reveals Regulation of the Ancient Kinase-like UbiB Family.
Cell chemical biology (2017-12-05)
Andrew G Reidenbach, Zachary A Kemmerer, Deniz Aydin, Adam Jochem, Molly T McDevitt, Paul D Hutchins, Jaime L Stark, Jonathan A Stefely, Thiru Reddy, Alex S Hebert, Emily M Wilkerson, Isabel E Johnson, Craig A Bingman, John L Markley, Joshua J Coon, Matteo Dal Peraro, David J Pagliarini
ABSTRACT
Human COQ8A (ADCK3) and Saccharomyces cerevisiae Coq8p (collectively COQ8) are UbiB family proteins essential for mitochondrial coenzyme Q (CoQ) biosynthesis. However, the biochemical activity of COQ8 and its direct role in CoQ production remain unclear, in part due to lack of known endogenous regulators of COQ8 function and of effective small molecules for probing its activity in vivo. Here, we demonstrate that COQ8 possesses evolutionarily conserved ATPase activity that is activated by binding to membranes containing cardiolipin and by phenolic compounds that resemble CoQ pathway intermediates. We further create an analog-sensitive version of Coq8p and reveal that acute chemical inhibition of its endogenous activity in yeast is sufficient to cause respiratory deficiency concomitant with CoQ depletion. Collectively, this work defines lipid and small-molecule modulators of an ancient family of atypical kinase-like proteins and establishes a chemical genetic system for further exploring the mechanistic role of COQ8 in CoQ biosynthesis.
Sigma-Aldrich
Triton X-100 reduced
Avanti
18:1 CDP DG, Avanti Research - A Croda Brand
Avanti
18:1 CDP DG, Avanti Research - A Croda Brand 870520C