Merck
  • Water and solute permeability of rat lung caveolae: high permeabilities explained by acyl chain unsaturation.
Water and solute permeability of rat lung caveolae: high permeabilities explained by acyl chain unsaturation.
American journal of physiology. Cell physiology (2005-02-25)
Warren G Hill, Eyad Almasri, W Giovanni Ruiz, Gerard Apodaca, Mark L Zeidel
ABSTRACT
Caveolae are invaginated membrane structures with high levels of cholesterol, sphingomyelin, and caveolin protein that are predicted to exist as liquid-ordered domains with low water permeability. We isolated a caveolae-enriched membrane fraction without detergents from rat lung and characterized its permeability properties to nonelectrolytes and protons. Membrane permeability to water was 2.85 +/- 0.41 x 10(-3) cm/s, a value 5-10 times higher than expected based on comparisons with other cholesterol and sphingolipid-enriched membranes. Permeabilities to urea, ammonia, and protons were measured and found to be moderately high for urea and ammonia at 8.85 +/- 2.40 x 10(-7)and 6.84 +/- 1.03 x 10(-2) respectively and high for protons at 8.84 +/- 3.06 x 10(-2) cm/s. To examine whether caveolin or other integral membrane proteins were responsible for high permeabilities, liposomes designed to mimic the lipids of the inner and outer leaflets of the caveolar membrane were made. Osmotic water permeability to both liposome compositions were determined and a combined inner/outer leaflet water permeability was calculated and found to be close to that of native caveolae at 1.58 +/- 1.1 x 10(-3) cm/s. In caveolae, activation energy for water flux was high (19.4 kcal/mol) and water permeability was not inhibited by HgCl2; however, aquaporin 1 was detectable by immunoblotting. Immunostaining of rat lung with AQP1 and caveolin antisera revealed very low levels of colocalization. We conclude that aquaporin water channels do not contribute significantly to the observed water flux and that caveolae have relatively high water and solute permeabilities due to the high degree of unsaturation in their fatty acyl chains.
Product Description
Avanti
18:0-18:1 PE, 1-stearoyl-2-oleoyl-sn-glycero-3-phosphoethanolamine, powder
Avanti
18:0-18:2 PS, 1-stearoyl-2-linoleoyl-sn-glycero-3-phospho-L-serine (sodium salt), chloroform
Avanti
18:0-18:2 PS, 1-stearoyl-2-linoleoyl-sn-glycero-3-phospho-L-serine (sodium salt), powder
Avanti
16:0-18:1 PC, Avanti Research - A Croda Brand
Avanti
16:0-18:1 PC, Avanti Research - A Croda Brand
Avanti
16:0-18:1 PA, powder
Avanti
Soy PI, Avanti Research - A Croda Brand 840044C
Avanti
Soy PI, Avanti Research - A Croda Brand
Avanti
18:0-20:4 PE, 1-stearoyl-2-arachidonoyl-sn-glycero-3-phosphoethanolamine, chloroform
Avanti
16:0 PC (DPPC), Avanti Research - A Croda Brand
Avanti
16:0-18:1 PA, 1-palmitoyl-2-oleoyl-sn-glycero-3-phosphate (sodium salt), chloroform
Avanti
16:0 PC (DPPC), Avanti Research - A Croda Brand
Avanti
18:1 (Δ9-Cis) PC (DOPC), Avanti Research - A Croda Brand
Avanti
18:1 (Δ9-Cis) PC (DOPC), Avanti Research - A Croda Brand
Avanti
Brain SM, Avanti Research - A Croda Brand
Avanti
18:0-22:6 PE, 1-stearoyl-2-docosahexaenoyl-sn-glycero-3-phosphoethanolamine, chloroform
Avanti
Brain SM, Avanti Research - A Croda Brand
Avanti
18:0-20:4 PC, 1-stearoyl-2-arachidonoyl-sn-glycero-3-phosphocholine, chloroform