Merck
  • Discovery of novel cyanodihydropyridines as potent mineralocorticoid receptor antagonists.
Discovery of novel cyanodihydropyridines as potent mineralocorticoid receptor antagonists.
Journal of medicinal chemistry (2010-08-03)
Graciela B Arhancet, Scott S Woodard, Kaliappan Iyanar, Brenda L Case, Rhonda Woerndle, Jessica D Dietz, Danny J Garland, Joe T Collins, Maria A Payne, James R Blinn, Silvia I Pomposiello, Xiao Hu, Marcia I Heron, Horng-Chih Huang, Len F Lee
ABSTRACT
A new 1,4-dihydropyridine 5a, containing a cyano group at the C3 position, was recently reported to possess excellent mineralocorticoid receptor (MR) antagonist in vitro potency and no calcium channel-blocker (CCB) activity. In the present study, we report the structure-activity relationships of this novel series of cyano ester dihydropyridines that resulted in R6 substituted analogues with improved metabolic stability while maintaining excellent MR antagonist activity and selectivity against other nuclear receptors. Further structure optimization with the introduction of five-membered ring heterocycles at R6 resulted in compounds with excellent MR antagonist potency and a suitable pharmacokinetic profile. In vivo studies of a promising tool compound in the Dahl salt-sensitive rat model of hypertension showed similar blood pressure (BP) reduction as the steroidal MR antagonist eplerenone, providing proof-of-concept (POC) for a nonsteroidal, orally efficacious MR antagonist.
Product Description
Supelco
Discovery® Cyano HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4.6 mm
Supelco
Discovery® Cyano HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4 mm
Supelco
Discovery® Cyano HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
Supelco
Discovery® Cyano Supelguard Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm