Merck
  • A phosphatidylinositol 4,5-bisphosphate redistribution-based sensing mechanism initiates a phagocytosis programing.
A phosphatidylinositol 4,5-bisphosphate redistribution-based sensing mechanism initiates a phagocytosis programing.
Nature communications (2018-10-17)
Libing Mu, Zhongyuan Tu, Lin Miao, Hefei Ruan, Ning Kang, Yongzhen Hei, Jiahuan Chen, Wei Wei, Fangling Gong, Bingjie Wang, Yanan Du, Guanghui Ma, Matthias W Amerein, Tie Xia, Yan Shi
ABSTRACT
Phagocytosis is one of the earliest cellular functions, developing approximately 2 billion years ago. Although FcR-based phagocytic signaling is well-studied, how it originated from ancient phagocytosis is unknown. Lipid redistribution upregulates a phagocytic program recapitulating FcR-based phagocytosis with complete dependence on Src family kinases, Syk, and phosphoinositide 3-kinases (PI3K). Here we show that in phagocytes, an atypical ITAM sequence in the ancient membrane anchor protein Moesin transduces signal without receptor activation. Plasma membrane deformation created by solid structure binding generates phosphatidylinositol 4,5-bisphosphate (PIP2) accumulation at the contact site, which binds the Moesin FERM domain and relocalizes Syk to the membrane via the ITAM motif. Phylogenic analysis traces this signaling using PI3K and Syk to 0.8 billion years ago, earlier than immune receptor signaling. The proposed general model of solid structure phagocytosis implies a preexisting lipid redistribution-based activation platform collecting intracellular signaling components for the emergence of immune receptors.
Product Description
Avanti
TopFluor TMR PC, Avanti Research - A Croda Brand 810180C
Avanti
TopFluor TMR PC, Avanti Research - A Croda Brand
Sigma-Aldrich
Uric acid, ≥99%, crystalline
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Wortmannin, InSolution, ≥95%, fungal metabolite that acts as a selective, cell-permeable and irreversible inhibitor of phosphatidylinositol 3-kinase (PI 3-kinase)
Sigma-Aldrich
Syk Inhibitor IV, BAY 61-3606, The Syk Inhibitor IV, BAY 61-3606, also referenced under CAS 732938-37-8, controls the biological activity of Syk. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.
Sigma-Aldrich
LY 294002, InSolution, ≥98%, 10 mM, reversible and specific inhibitor of PI 3-kinase
Sigma-Aldrich
Syk Inhibitor, The Spleen Tyrosine Kinase Inhibitor, also referenced under CAS 622387-85-3, controls the biological activity of Spleen Tyrosine Kinase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.
Sigma-Aldrich
Piceatannol, powder
Sigma-Aldrich
Piceatannol, A cell-permeable, substrate competitive and reversible plant metabolite that inhibits the activity of rat liver protein kinase A catalytic subunit (IC₅₀ = 3 µM), PKC (IC₅₀ = 8 µM), MLCK (IC₅₀ = 12 µM).