Merck
FABP4 as a key determinant of metastatic potential of ovarian cancer.
Nature communications (2018-07-28)
Kshipra M Gharpure, Sunila Pradeep, Marta Sans, Rajesha Rupaimoole, Cristina Ivan, Sherry Y Wu, Emine Bayraktar, Archana S Nagaraja, Lingegowda S Mangala, Xinna Zhang, Monika Haemmerle, Wei Hu, Cristian Rodriguez-Aguayo, Michael McGuire, Celia Sze Ling Mak, Xiuhui Chen, Michelle A Tran, Alejandro Villar-Prados, Guillermo Armaiz Pena, Ragini Kondetimmanahalli, Ryan Nini, Pranavi Koppula, Prahlad Ram, Jinsong Liu, Gabriel Lopez-Berestein, Keith Baggerly, Livia S Eberlin, Anil K Sood
ABSTRACT
The standard treatment for high-grade serous ovarian cancer is primary debulking surgery followed by chemotherapy. The extent of metastasis and invasive potential of lesions can influence the outcome of these primary surgeries. Here, we explored the underlying mechanisms that could increase metastatic potential in ovarian cancer. We discovered that FABP4 (fatty acid binding protein) can substantially increase the metastatic potential of cancer cells. We also found that miR-409-3p regulates FABP4 in ovarian cancer cells and that hypoxia decreases miR-409-3p levels. Treatment with DOPC nanoliposomes containing either miR-409-3p mimic or FABP4 siRNA inhibited tumor progression in mouse models. With RPPA and metabolite arrays, we found that FABP4 regulates pathways associated with metastasis and affects metabolic pathways in ovarian cancer cells. Collectively, these findings demonstrate that FABP4 is functionally responsible for aggressive patterns of disease that likely contribute to poor prognosis in ovarian cancer.
Sigma-Aldrich
Monoclonal Anti-Vinculin antibody produced in mouse, clone hVIN-1, ascites fluid
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid
Sigma-Aldrich
Anti-FABP4 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution