Merck
  • Morpholine catalyzed direct C3 alkenylation of indoles with α,β-unsaturated aldehydes.
Morpholine catalyzed direct C3 alkenylation of indoles with α,β-unsaturated aldehydes.
Chemical communications (Cambridge, England) (2011-06-21)
Shi-Kai Xiang, Bo Zhang, Li-He Zhang, Yuxin Cui, Ning Jiao
ABSTRACT
Organocatalytic direct C3 alkenylation of indoles has been developed. Simple and readily available morpholine trifluoroacetic acid salt is employed as an efficient catalyst in this oxidative dehydrogenative reaction. Simplicity and practicality constitute the most attractive advantages of this reaction.
Product Description
Supelco
Morpholine, analytical standard
Sigma-Aldrich
Morpholine, ACS reagent, ≥99.0%
Sigma-Aldrich
Morpholine, purified by redistillation, ≥99.5%
Sigma-Aldrich
Morpholine, ReagentPlus®, ≥99%