Skip to Content
Merck
(Vinylbenzyl)trimethylammonium chloride 99%

(Vinylbenzyl)trimethylammonium chloride

(Vinylbenzyl)trimethylammonium chloride 99%
Synonym(s):
(ar-Vinylbenzyl)trimethylammonium chloride
Linear Formula:
H2C=CHC6H4CH2N(CH3)3Cl
CAS Number:
Molecular Weight:
211.73
EC Number:

Products

Brand
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Markush Class
Markush Group
Monomer Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing