
Triadimefon

Synonym(s):
1-(4-Chlorophenoxy)-3,3-dimethyl-1-(1H-1,2,4-triazol-1-yl)-2-butanone
Empirical Formula (Hill Notation):
C14H16ClN3O2
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2