
Tolnaftate

Synonym(s):
Methyl-(3-methylphenyl)carbamothioic acid O-2-naphthyl ester, Tolnaftate
Empirical Formula (Hill Notation):
C19H17NOS
CAS Number:
Molecular Weight:
307.41
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3