
Tigecycline Related Compound B

Synonym(s):
9-Aminominocycline hydrochloride, (4S,4aS,5aR,12aS)-9-Amino-4,7-bis(dimethylamino)-3,10,12,12a-tetrahydroxy-1,11-dioxo-1,4,4a,5,5a,6,11,12a-octahydrotetracene-2-carboxamide hydrochloride
Empirical Formula (Hill Notation):
C23H28N4O7 · HCl
CAS Number:
Molecular Weight:
508.95
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2