
Teflubenzuron

Empirical Formula (Hill Notation):
C14H6Cl2F4N2O2
CAS Number:
Molecular Weight:
381.11
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-2 of 2